Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%

Supplier: AMBEED, INC
Warning

A187462-10G A187462-1G A187462-250MG A187462-5G
76769-798EA 120 USD InStock
76769-798 76769-800 76769-802 77625-898
5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%
5-Bromo-2-chloro-4-(methylthio)pyrimidine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂S
MDL Number: MFCD07781218
CAS Number: 59549-51-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty