Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isopropylphthalimide 97%

Supplier: AMBEED, INC
Warning

A193459-25G A193459-5G
76769-942EA 87 USD InStock
76769-942 76769-944
N-Isopropylphthalimide 97%
N-Isopropylphthalimide
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MDL Number: MFCD00014582
CAS Number: 304-17-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty