Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutylamine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A196286-10G A196286-1G
76769-982EA 50 USD InStock
76769-982 76769-984
Cyclobutylamine hydrochloride 97%
Cyclobutylamine hydrochloride


Formula: C₄H₉N·HCl
MDL Number: MFCD00034953
CAS Number: 6291-01-6
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty