Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 4-vinylbenzenesulfonate hydrate 98%

Supplier: AMBEED, INC
Warning

A196604-5G A196604-25G
76769-988EA 5 USD InStock
76769-988 77302-520
Sodium 4-vinylbenzenesulfonate hydrate 98%
Sodium 4-vinylbenzenesulfonate hydrate
Precaución: for Research Use Only


Formula: C₈H₉NaO₄S
MW: 224.20 g/mol
MDL Number: MFCD03092905
CAS Number: 304675-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty