Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-fluoro-2-methylaniline 98%

Supplier: AMBEED, INC
Warning

A197024-10G A197024-1G A197024-25G A197024-5G
76770-008EA 20 USD InStock
76770-008 76770-010 76770-012 76770-014
4-Bromo-5-fluoro-2-methylaniline 98%
4-Bromo-5-fluoro-2-methylaniline
Precaución: for Research Use Only


Formula: C₇H₇BrFN
MW: 204.0400 g/mol
MDL Number: MFCD04973753
CAS Number: 52723-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty