Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate 97%

Supplier: AMBEED, INC
Warning

A198872-100MG A198872-1G A198872-250MG
76770-076EA 96 USD InStock
76770-076 76770-078 76770-080
tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate 97%
tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD09991605
CAS Number: 122848-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty