Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-tert-Butyl-2-hydroxyisophthalaldehyde 95%

Supplier: AMBEED, INC
Warning

A199557-10G A199557-1G A199557-5G
76770-100EA 216 USD InStock
76770-100 76770-102 76770-104
5-tert-Butyl-2-hydroxyisophthalaldehyde 95%
5-tert-Butyl-2-hydroxyisophthalaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
MDL Number: MFCD01567135
CAS Number: 84501-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty