Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)benzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A200632-1G A200632-25G A200632-5G
76770-130EA 29.71 USD InStock
76770-130 76770-132 76770-134 76770-136
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride 98%
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₈H₃ClF₆O₂S
MW: 312.62
MDL Number: MFCD00014725
CAS Number: 39234-86-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty