Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluorogramine 98%

Supplier: AMBEED, INC
Warning

A202511-1G A202511-250MG A202511-25G A202511-5G
76770-206EA 18 USD InStock
76770-206 76770-208 76770-210 76770-212
5-Fluorogramine 98%
5-Fluorogramine
Precaución: for Research Use Only


Formula: C₁₁H₁₃FN₂
MDL Number: MFCD00022731
CAS Number: 343-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty