Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitrotoluene 98%

Supplier: AMBEED, INC
Warning

A204068-100G A204068-10G A204068-1G A204068-25G A204068-500G A204068-5G
76770-254EA 26 USD InStock
76770-254 76770-256 76770-258 76770-260 76770-262 76770-264
5-Fluoro-2-nitrotoluene 98%
5-Fluoro-2-nitrotoluene
Precaución: for Research Use Only


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 218
Melting Pt: 28
MDL Number: MFCD00007283
CAS Number: 446-33-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty