Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimorpholinophosphine oxide 97%

Supplier: AMBEED, INC
Warning

A209003-100G A209003-25G A209003-5G
76770-408EA 203 USD InStock
76770-408 76770-410 76770-412 76770-414
Trimorpholinophosphine oxide 97%
Trimorpholinophosphine oxide
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₃O₄P
MDL Number: MFCD00047406
CAS Number: 4441-12-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty