Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodanine-N-propionic acid 97%

Supplier: AMBEED, INC
Warning

A209870-100G A209870-1G A209870-250MG A209870-5G
76770-456EA 335 USD InStock
76770-456 76770-458 76770-460 76770-462
Rhodanine-N-propionic acid 97%
Rhodanine-N-propionic acid
Precaución: for Research Use Only


Formula: C₆H₇NO₃S₂
MDL Number: MFCD00022558
CAS Number: 7025-19-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty