Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-fluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A211584-100G A211584-10G A211584-5G
76770-538EA 77 USD InStock
76770-538 76770-540 76770-542
3-Bromo-4-fluorobenzonitrile 98%
3-Bromo-4-fluorobenzonitrile
Precaución: For research use only.


Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD00055432
CAS Number: 79630-23-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty