Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylphenylhydrazinium chloride 95%

Supplier: AMBEED, INC
Warning

A212016-10G A212016-25G A212016-5G
76770-552EA 15 USD InStock
76770-552 76770-554 76770-556
3,5-Dimethylphenylhydrazinium chloride 95%
3,5-Dimethylphenylhydrazinium chloride
Precaución: for Research Use Only


Formula: C₈H₁₃ClN₂
MDL Number: MFCD00052269
CAS Number: 60481-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty