Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(hydroxymethyl)phenol 95%

Supplier: AMBEED, INC
Warning

A212095-1G A212095-250MG
76770-558EA 39 USD InStock
76770-558 76770-560
5-Bromo-2-(hydroxymethyl)phenol 95%
5-Bromo-2-(hydroxymethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₇BrO₂
MW: 203.04 g/mol
MDL Number: MFCD08445665
CAS Number: 170434-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty