Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyisoindolin-1-one 97%

Supplier: AMBEED, INC
Warning

A212875-10G A212875-1G A212875-250MG A212875-5G
76770-594EA 358 USD InStock
76770-594 76770-596 76770-598 76770-600
6-Methoxyisoindolin-1-one 97%
6-Methoxyisoindolin-1-one
Precaución: for Research Use Only


Formula: C₉H₉NO₂
MDL Number: MFCD08234735
CAS Number: 132680-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty