Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-nitropyridine-2-carboxamide 95%

Supplier: AMBEED, INC
Warning

A216688-100MG A216688-10G A216688-1G A216688-250MG
76770-700EA 33 USD InStock
76770-700 76770-702 76770-704 76770-706
6-Chloro-3-nitropyridine-2-carboxamide 95%
6-Chloro-3-nitropyridine-2-carboxamide
Precaución: for Research Use Only


Formula: C₆H₄ClN₃O₃
MDL Number: MFCD07780776
CAS Number: 171178-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty