Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Benzyloctahydropyrrolo[3,4-b]pyridine 96%

Supplier: AMBEED, INC
Warning

A222096-250MG A222096-5G
76770-860EA 34 USD InStock
76770-860 77626-116
6-Benzyloctahydropyrrolo[3,4-b]pyridine 96%
6-Benzyloctahydropyrrolo[3,4-b]pyridine
Precaución: for Research Use Only


Formula: C₁₄H₂₀N₂
MDL Number: MFCD06796542
CAS Number: 128740-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty