Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-methyl-1H-benzo[d]imidazole 95%

Supplier: AMBEED, INC
Warning

A222376-1G A222376-250MG A222376-25G A222376-5G
76770-870EA 38 USD InStock
76770-870 76770-872 76770-874 76770-876
6-Chloro-4-methyl-1H-benzo[d]imidazole 95%
6-Chloro-4-methyl-1H-benzo[d]imidazole
Precaución: for Research Use Only


Formula: C₈H₇ClN₂
MDL Number: MFCD02181079
CAS Number: 180508-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty