Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-6-methylpyrimidine-2,4-diol 97%

Supplier: AMBEED, INC
Warning

A226539-10G A226539-1G A226539-250MG A226539-25G A226539-5G
76771-006EA 47 USD InStock
76771-006 76771-008 76771-010 76771-012 76771-014
5-Amino-6-methylpyrimidine-2,4-diol 97%
5-Amino-6-methylpyrimidine-2,4-diol
Precaución: for Research Use Only


Formula: C₅H₇N₃O₂
MDL Number: MFCD00053572
CAS Number: 6270-46-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty