Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-amino-2-thioxo-1,2-dihydropyrimidine-5-carboxylate 98%

Supplier: AMBEED, INC
Warning

A229979-1G A229979-25G A229979-5G
76771-118EA 15 USD InStock
76771-118 76771-120 76771-122
Ethyl 6-amino-2-thioxo-1,2-dihydropyrimidine-5-carboxylate 98%
Ethyl 6-amino-2-thioxo-1,2-dihydropyrimidine-5-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉N₃O₂S
MDL Number: MFCD00614345
CAS Number: 774-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty