Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butyl-o-xylene 99+%

Supplier: AMBEED, INC
Warning

A234876-100G A234876-25G A234876-5G
76771-230EA 123 USD InStock
76771-230 76771-232 76771-234
4-tert-Butyl-o-xylene 99+%
4-tert-Butyl-o-xylene
Precaución: for Research Use Only


Formula: C₁₂H₁₈
MW: 162.27 g/mol
MDL Number: MFCD00038277
CAS Number: 7397-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty