Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A236089-100G A236089-10G A236089-25G A236089-500G A236089-5G
76771-274EA 55 USD InStock
76771-274 76771-276 76771-278 76771-280 76771-282
6-Chloro-2-hydroxypyridine 98%
6-Chloro-2-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD00006278
CAS Number: 16879-02-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty