Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-6-chloro-5-methylpyrimidine 95%

Supplier: AMBEED, INC
Warning

A237075-100MG A237075-1G A237075-250MG A237075-5G
76771-298EA 11 USD InStock
76771-298 76771-300 76771-302 76771-304
4-Amino-6-chloro-5-methylpyrimidine 95%
4-Amino-6-chloro-5-methylpyrimidine
Precaución: for Research Use Only


Formula: C₅H₆ClN₃
MDL Number: MFCD09991585
CAS Number: 14394-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty