Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A237408-100MG A237408-250MG A237408-1G
76771-322EA 129 USD InStock
76771-322 76771-324 77626-264
6-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClF₃N
MDL Number: MFCD08234660
CAS Number: 215798-14-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty