Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2-methylbenzothiazole 98%

Supplier: AMBEED, INC
Warning

A243855-1G A243855-250MG A243855-25G A243855-5G
76771-464EA 20 USD InStock
76771-464 76771-466 76771-468 76771-470
6-Methoxy-2-methylbenzothiazole 98%
6-Methoxy-2-methylbenzothiazole
Precaución: for Research Use Only


Formula: C₉H₉NOS
MDL Number: MFCD00005795
CAS Number: 2941-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty