Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluorobenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A250079-100G A250079-10G A250079-1G A250079-25G A250079-500G A250079-5G
76771-622EA 119 USD InStock
76771-622 76771-624 76771-626 76771-628 76771-630 76771-632
2-Chloro-5-fluorobenzaldehyde 95%
2-Chloro-5-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD03788511
CAS Number: 84194-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty