Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-amino-1,3-thiazole-5-carboxylate 95%

Supplier: AMBEED, INC
Warning

A252490-250MG A252490-1G A252490-100MG
76771-684EA 17 USD InStock
76771-684 77593-586 77626-326
Methyl 4-amino-1,3-thiazole-5-carboxylate 95%
Methyl 4-amino-1,3-thiazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₅H₆N₂O₂S
MW: 158.178 g/mol
MDL Number: MFCD09033876
CAS Number: 278183-10-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty