Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monosodium 5-sulfoisophthalate 97%

Supplier: AMBEED, INC
Warning

A252507-25G A252507-100G
76771-686EA 5 USD InStock
76771-686 77626-328
Monosodium 5-sulfoisophthalate 97%
Monosodium 5-sulfoisophthalate
Precaución: for Research Use Only


Formula: C₈H₆O₇S
MDL Number: MFCD00007495
CAS Number: 6362-79-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty