Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-hydroxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A253957-100G A253957-10G A253957-1G A253957-25G A253957-500G A253957-5G
76771-738EA 223 USD InStock
76771-738 76771-740 76771-742 76771-744 76771-746 76771-748
2-Fluoro-4-hydroxybenzaldehyde 98%
2-Fluoro-4-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅FO₂
MDL Number: MFCD06797918
CAS Number: 348-27-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty