Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethylpicolinic acid 98%

Supplier: AMBEED, INC
Warning

A261121-100MG A261121-1G A261121-250MG
76771-868EA 41 USD InStock
76771-868 76771-870 76771-872
5-Ethylpicolinic acid 98%
5-Ethylpicolinic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00995160
CAS Number: 770-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty