Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-methylsalicylate 98%

Supplier: AMBEED, INC
Warning

A261354-100G A261354-10G A261354-250MG A261354-25G A261354-5G
76771-880EA 307 USD InStock
76771-880 76771-882 76771-884 76771-886 76771-888
Ethyl-6-methylsalicylate 98%
Ethyl-6-methylsalicylate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD01194299
CAS Number: 6555-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty