Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(benzylamino)propionate 97%

Supplier: AMBEED, INC
Warning

A262660-100G A262660-10G A262660-1G A262660-25G
76771-906EA 95 USD InStock
76771-906 76771-908 76771-910 76771-912
Ethyl-3-(benzylamino)propionate 97%
Ethyl-3-(benzylamino)propionate


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD00085541
CAS Number: 23583-21-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty