Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A263664-100G A263664-10G A263664-25G A263664-500G A263664-5G
76771-934EA 115 USD InStock
76771-934 76771-936 76771-938 76771-940 76771-942
3-Chloro-2-fluorobenzonitrile 98%
3-Chloro-2-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClFN
MW: 155.56
MDL Number: MFCD00042206
CAS Number: 94087-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty