Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-3-boronic acid neopentylglycol ester 95%

Supplier: AMBEED, INC
Warning

A264751-10G A264751-1G A264751-5G
76771-972EA 83 USD InStock
76771-972 76771-974 76771-976
Pyridine-3-boronic acid neopentylglycol ester 95%
Pyridine-3-boronic acid neopentylglycol ester
Precaución: for Research Use Only


Formula: C₁₀H₁₄BNO₂
MDL Number: MFCD03425979
CAS Number: 845885-86-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty