Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethyl-2-naphthalenol 98%

Supplier: AMBEED, INC
Warning

A264852-100MG A264852-1G A264852-250MG A264852-5G
76771-978EA 35 USD InStock
76771-978 76771-980 76771-982 76771-984
6-Ethyl-2-naphthalenol 98%
6-Ethyl-2-naphthalenol
Precaución: for Research Use Only


Formula: C₁₂H₁₂O
MDL Number: MFCD07368645
CAS Number: 1999-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty