Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

YM-90709 98%

Supplier: AMBEED, INC
Warning

A265624-10MG A265624-1MG A265624-25MG A265624-5MG
76771-998EA 35 USD InStock
76771-998 76772-000 76772-002 76772-004
YM-90709 98%
YM-90709
Precaución: for Research Use Only


Formula: C₂₂H₂₁N₃O₂
MW: 359.43 g/mol
MDL Number: MFCD00354370
CAS Number: 163769-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty