Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluorobenzyl alcohol 95%

Supplier: AMBEED, INC
Warning

A267150-100G A267150-25G A267150-5G
76772-050EA 88 USD InStock
76772-050 76772-052 76772-054
2-Chloro-6-fluorobenzyl alcohol 95%
2-Chloro-6-fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 232
MDL Number: MFCD00004608
CAS Number: 56456-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty