Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-biphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A267411-100G A267411-25G A267411-5G
76772-068EA 349 USD InStock
76772-068 76772-070 76772-072
2-Fluoro-4-biphenylboronic acid 98%
2-Fluoro-4-biphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀BFO₂
MW: 216.02 g/mol
MDL Number: MFCD01075707
CAS Number: 178305-99-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty