Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-1H-1,2,3-benzotriazole 95%

Supplier: AMBEED, INC
Warning

A270780-100MG A270780-1G A270780-250MG A270780-5G
76772-140EA 10 USD InStock
76772-140 76772-142 76772-144 76772-146
5-Fluoro-1H-1,2,3-benzotriazole 95%
5-Fluoro-1H-1,2,3-benzotriazole
Precaución: for Research Use Only


Formula: C₆H₄FN₃
MDL Number: MFCD00760403
CAS Number: 18225-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty