Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-methylbenzo[d]thiazole 98%

Supplier: AMBEED, INC
Warning

A274246-1G A274246-250MG A274246-25G A274246-5G
76772-196EA 18 USD InStock
76772-196 76772-198 76772-200 76772-202
6-Bromo-2-methylbenzo[d]thiazole 98%
6-Bromo-2-methylbenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₈H₆BrNS
MW: 228.1089 g/mol
MDL Number: MFCD00466580
CAS Number: 5304-21-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty