Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloroisatin 98%

Supplier: AMBEED, INC
Warning

A277595-100G A277595-25G A277595-500G A277595-5G
76772-256EA 59 USD InStock
76772-256 76772-258 76772-260 76772-262
5-Chloroisatin 98%
5-Chloroisatin
Precaución: for Research Use Only


Formula: C₈H₄ClNO₂
MDL Number: MFCD00014567
CAS Number: 17630-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty