Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)-2-chloroaniline 98%

Supplier: AMBEED, INC
Warning

A279751-100G A279751-25G A279751-5G
76772-280EA 90 USD InStock
76772-280 76772-282 76772-284
3,5-Bis(trifluoromethyl)-2-chloroaniline 98%
3,5-Bis(trifluoromethyl)-2-chloroaniline
Precaución: for Research Use Only


Formula: C₈H₄ClF₆N
MDL Number: MFCD04038716
CAS Number: 201593-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty