Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,4-dimethylnicotinate 98%

Supplier: AMBEED, INC
Warning

A281556-10G A281556-1G A281556-25G A281556-5G
76772-304EA 117 USD InStock
76772-304 76772-306 76772-308 76772-310
Ethyl-2,4-dimethylnicotinate 98%
Ethyl-2,4-dimethylnicotinate
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00051962
CAS Number: 37669-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty