Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-4-octanone 85% (by GC)

Supplier: AMBEED, INC
Warning

A284008-100G A284008-25G A284008-500G
76772-364EA 274 USD InStock
76772-364 76772-366 76772-368
5-Hydroxy-4-octanone 85% (by GC)
5-Hydroxy-4-octanone
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 188
Melting Pt: 2
MDL Number: MFCD00021928
CAS Number: 496-77-5
Merck Index: 13,01595
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty