Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A305015-100G A305015-10G A305015-25G A305015-5G
76772-692EA 46 USD InStock
76772-692 76772-694 76772-696 76772-698
4-Fluoro-2-(trifluoromethyl)aniline 98%
4-Fluoro-2-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅F₄N
MDL Number: MFCD00007831
CAS Number: 393-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty