Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3-nitro-p-toluic acid 98%

Supplier: AMBEED, INC
Danger

A308455-10G A308455-1G A308455-5G
76772-746EA 144 USD InStock
76772-746 76772-748 76772-750
α-Bromo-3-nitro-p-toluic acid 98%
α-Bromo-3-nitro-p-toluic acid
Precaución: for Research Use Only


Formula: C₈H₆BrNO₄
MW: 260.04 g/mol
Melting Pt: 132
MDL Number: MFCD00216575
CAS Number: 55715-03-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty