Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Dichloro-2,3-dicyanopyrazine 96%

Supplier: AMBEED, INC
Danger

A310388-10G A310388-1G A310388-5G
76772-760EA 45 USD InStock
76772-760 76772-762 76772-764 76772-766
5,6-Dichloro-2,3-dicyanopyrazine 96%
5,6-Dichloro-2,3-dicyanopyrazine
Precaución: for Research Use Only


Formula: C₆Cl₂N₄
MDL Number: MFCD00191419
CAS Number: 56413-95-7
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty