Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluorobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A318013-100G A318013-10G A318013-25G A318013-5G
76772-878EA 60 USD InStock
76772-878 76772-880 76772-882 76772-884
2,5-Difluorobenzoic acid 98%
2,5-Difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 242
MDL Number: MFCD00002410
CAS Number: 2991-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty