Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-1H-indazol-5-amine 95%

Supplier: AMBEED, INC
Warning

A327742-100MG A327742-1G A327742-250MG
76773-044EA 14 USD InStock
76773-044 76773-046 76773-048
6-Methyl-1H-indazol-5-amine 95%
6-Methyl-1H-indazol-5-amine
Precaución: for Research Use Only


Formula: C₈H₉N₃
MDL Number: MFCD03305372
CAS Number: 81115-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty